missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Brilliant Green
Description
Brilliant Green a stain used in bacteriological media.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Brilliant Green a stain used in bacteriological media.
Solubility
Soluble in water (100 mg/ml at 25°C), ethanol (50 mg/ml), methanol, and 2-methoxyethanol (60 mg/ml).
Notes
Store away from oxidizing agents. Stable under recommended storage conditions.
Brilliant Green a stain used in bacteriological media.
Solubility
Soluble in water (100 mg/ml at 25°C), ethanol (50 mg/ml), methanol, and 2-methoxyethanol (60 mg/ml).
Notes
Store away from oxidizing agents. Stable under recommended storage conditions.
Specifications
Specifications
| Melting Point | 210°C (decomposition) |
| Beilstein | 3901207 |
| Merck Index | 14,1374 |
| Solubility Information | Soluble in water (100mg/ml at 25°C),ethanol (50mg/ml),methanol,and 2-methoxyethanol (60mg/ml). |
| Formula Weight | 482.65 |
| Quantity | 25 g |
| Chemical Name or Material | Brilliant Green |
Safety and Handling
| Hazard Category | H302-H319 |
| Hazard Statement | GHS H Statement H302-H315-H319-H335 Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c |
| EINECSNumber | 211-190-1 |
| RTECSNumber | BP6825000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only