missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Brilliant cresyl Blue, pure, high purity biological stain

CAS: 81029-05-2 | 0·5 ZnCl4

$197.77 - $413.65

Chemical Identifiers

CAS 81029-05-2
Molecular Formula 0·5 ZnCl4
MDL Number MFCD00148902
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC229590100
SDS View Documents
Thermo Scientific Chemicals
229590100
10 g Glass bottle
Each for $197.77
Only null left
 
AC229590250
SDS View Documents
Thermo Scientific Chemicals
229590250
25 g Glass bottle
Each for $413.65
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

81029-05-2
0·5 ZnCl4
MFCD00148902

Specifications

1500 (625 to 630nm, 50% min. ethanol) (A (1% min./1cm))
0·5 ZnCl4
Authentic
400.97
Glass bottle
White
Crystalline Powder
81029-05-2
MFCD00148902
8 % max. (1 g, 110°C)
Pure
1500 to 1700 (622nm, 50% ethanol)
10 g
Brilliant cresyl blue
Videos
Safety and Handling

Safety and Handling

LIGHT SENSITIVE

SDS
Documents

Documents

RUO – Research Use Only