accessibility menu, dialog, popup

UserName

ClH3CNCH3NONH2NH2CH3

CAS RN 81029-05-2

IUPAC Name: 7,9-diamino-N,N-diethyl-6-methyl-5a,9a-dihydro-3H-phenoxazin-3-iminium chloride
Synonyms: brilliant cresyl blue
Molecular Weight (g/mol): 334.85
Molecular Formula: C17H23ClN4O
InChi Key: SHMCAAFKAHAURK-UHFFFAOYNA-M
SMILES: [Cl-].CC[N+](CC)=C1C=CC2=NC3C(OC2=C1)C(C)=C(N)C=C3N
Color 
  • (1)
  • (2)
Grade 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)