missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Brilliant cresyl Blue, pure, high purity biological stain
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Absorbance | 1500 (625 to 630nm, 50% min. ethanol) (A (1% min./1cm)) |
| Infrared Spectrum | Authentic |
| Loss on Drying | 8 % max. (1 g, 110°C) |
| Packaging | Glass bottle |
| Specific Extinction | 1500 to 1700 (622nm, 50% ethanol) |
| Color | White |
| Quantity | 10 g |
| Physical Form | Crystalline Powder |
| Formula Weight | 400.97 |
| Grade | Pure |
| Show More |
Safety and Handling
| Hazard Category | LIGHT SENSITIVE |
RUO – Research Use Only