missing translation for 'onlineSavingsMsg'
Learn More

Brilliant Cresyl Blue, MP Biomedicals™

Used in hematology for demonstrating and counting reticulocytes and platelets. Brilliant Cresyl Blue, MP Biomedicals is a supravital stain that is also used as a vital stain for demonstrating the mucous secretions of the marine coelomates.

Supplier:  MP Biomedicals Inc 0215051525

Catalog No. MP215051525


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

81029-05-2
Brilliant Blue C, Brilliant Cresyl Blue ALD
385.94
Powder
C17H20N3OCl. 1/2(ZnCl2)
Soluble in water, 1N Hydrochloric acid, or 50% ethanol.
25 g

Chemical Identifiers

81029-05-2
C17H20N3OCl. 1/2(ZnCl2)
Brilliant Blue C, Brilliant Cresyl Blue ALD
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : Room Temperature (15–30°C).

SDS
Documents

Documents

Promotions

Promotions