missing translation for 'onlineSavingsMsg'
Learn More

Brilliant Black BN, ≥95% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013717
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013717 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013717 Supplier Merck Emd Millipore Supplier No. 1122025MG

missing translation for 'validMainframeMsg'

Brilliant Black BN (BB) is a azo dye which is basically a tetra sodium salt. It is a synthetic dye mainly used as food additive. It is used as colouring agent to enhance the appearance. It is usually used in jellies, brown sauces and cake mixes.

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 2519-30-4
Physical Form Neat
Linear Formula C28H17N5 Na4O14S4
MDL Number MFCD00078980
Synonym Food Black 1
Type Analytical Standard
Quantity 25 mg
Formula Weight 867.68
Percent Purity ≥95% (HPLC)
Grade Analytical Standard