missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brij(R) S100, Spectrum™ Chemical
B1593, 9005-00-9
Chemical Identifiers
| CAS | 9005-00-9 |
|---|---|
| Molecular Formula | C20H41O2 |
| Molecular Weight (g/mol) | 313.55 |
| InChI Key | PSEGVHKUPXNGGJ-UHFFFAOYSA-N |
| IUPAC Name | [2-(octadecyloxy)ethyl]oxidanyl |
| SMILES | CCCCCCCCCCCCCCCCCCOCC[O] |
Description
Spectrum™ Chemical Brij S100 , Polyethylene (100) Stearyl Ether, functions as an aqueous dispersant and oil/water emulsifier.