missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eBioscience™ BrdU Staining Kit for Flow Cytometry FITC
Description
Includes
- BrdU
- DNase I
- Anti-BrdU FITC Antibody (clone BU20A)
- BrdU Staining Buffer Concentrate (4X)
- Fixation/Permeabilization Diluent
- Cycling cells are incubated with 5-bromo-2′-deoxyuridine (BrdU), a synthetic analog of thymidine which incorporates into newly synthesized genomic DNA during the S-phase of mitosis
- Following DNA denaturation, the cells are stained for BrdU incorporation along with any other cell surface and/or intracellular targets of interest
Specifications
Specifications
| Detection Method | Fluorescence |
| For Use With (Application) | BrdU Staining |
| For Use With (Equipment) | Flow Cytometer |
| Product Type | Staining Kit |
| Format | Kit |
| Product Line | eBioscience |
| Quantity | 100 Tests |
For Research Use Only. Not for use in diagnostic procedures.