missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ BrdU Kit for IHC/ICC Immunofluorescence eFluor™ 615

Catalog No. 501125105
Change view
Click to view available options
Quantity:
50 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501125105 50 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Contains Sodium azide and formaldehyde
Catalog No. 501125105 Supplier Invitrogen™ Supplier No. 8842659945

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Contains Sodium azide and formaldehyde

Includes

  • BrdU Fixation Buffer
  • BrdU Antigen Retrieval Solution (10X)
  • BrdU Blocking Solution
  • Anti-BrdU Biotin (clone BU20A) : 10 μg
  • Streptavidin for BrdU IHC/ICC eFluor 615 : 3 μg

Contains reagents and buffers to identify and examine proliferating cells by immunohistochemical or immunocytochemical analysis. Optimized for IHC with both frozen and formalin-fixed paraffin-embedded BrdU-labeled mouse intestine and ICC of BrdU-pulsed HeLa cells grown on culture slides.

Cells are labeled in vitro or in vivo with 5-bromo-2’-deoxyuridine (BrdU), a synthetic analog of thymidine, which is incorporated into DNA in place of thymidine during the S-phase of the cell cycle. Following fixation and antigen retrieval steps, cells or tissue sections are stained for BrdU incorporation and visualized using a streptavidin-conjugated fluorophore.

Specifications

Conjugate BrdU
Detection Method Fluorometric
Quantity 50 tests
Product Line eBioscience
For Use With (Application) ICC (Immunochemistry), IHC (Immunohistochemistry)
Product Type IHC/ICC BrdU Kit
Target Species All species

For Research Use Only. Not for use in diagnostic procedures.