missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BRD3 Polyclonal Antibody

Description
Immunogen sequence: MPQEEVELLP PAPKGKGRKP AAGAQSAGTQ QVAAVSSVSP ATPFQSVPPT VSQTPVIAAT PVPTITANVT SV Highest antigen sequence indentity to the following orthologs: Mouse - 94%, Rat - 90%.
This gene was identified based on its homology to the gene encoding the RING3 protein, a serine/threonine kinase. The gene localizes to 9q34, a region which contains several major histocompatibility complex (MHC) genes. The function of the encoded protein is not known.
Specifications
Specifications
| Antigen | BRD3 |
| Applications | ChIP Assay, Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.10 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | BRD3 |
| Gene Accession No. | Q15059 |
| Gene Alias | 2410084F24Rik; AW060456; BRD3; bromodomain containing 3; bromodomain containing 3; bromodomain-containing protein 3; bromodomain containing protein 3; bromodomain-containing 3; bromodomain-containing FSH-like protein FSRG2; Bromodomain-containing protein 3; bromodomain-containing protein 3 short isoform; Fsrg2; KIAA0043; ORFX; RING3L; RING3-like protein; RINGL3 |
| Gene Symbols | BRD3 |
| Show More |