missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRANDTECH™ VITLAB™ Wide-Mouth Reagent Bottles with Screw Caps
Description
- Wide mouth for easier access to contents
- Screw caps help protect agaist outside contamination and leakage
Specifications
Specifications
| Capacity | 8.45 oz., 250 mL |
| Capacity (Metric) | 250 mL |
| Capacity (English) | 8.45 oz. |
| Color | Transparent |
| Diameter (English) | 2.87 in. |
| Material | Polypropylene |
| Quantity | 20/Pk. |
| Sterility | Non-sterile |
| Closure Included | Yes |
| Graduated | No |
| Show More |