Learn More
BRANDTECH™ VITLAB™ Styrene Acrylonitrile Copolymer Graduated Pitchers
VITLAB™ SAN graduated pitchers are transparent with molded graduations for durability
Supplier:  BRANDTECH™ VITLAB™ 44391
 View more versions of this product
Catalog No. 14379281
Only null left
Explore available promotions
Description
Description
  • Light weight alternative to glass
Specifications
Specifications
67.68 oz., 2000 mL
67.68 oz.
5.90 in.
Non-sterile
Reusable
Wide
215 mm
5.9 in.
150 mm
2000 mL
Transparent
SAN
No
Yes
Round
Pouring Pitcher
8.46 in.
Not Autoclavable
Product Suggestions
Product Suggestions