Learn More
BrandTech™ BRAND™ Reagent Reservoirs
Constructed of heavy-gauge polypropylene for long-lasting durability. Reagent Reservoirs are autoclavable and suitable for all 8- and 12-channel pipetters. Non-sterile version includes lid.
Supplier:  BrandTech™ BRAND™ 703411
 View more versions of this product
Catalog No. 13-689-208
Only null left
Explore available promotions
Description
Description
  • Heavy-gauge polypropylene
  • Suitable for all 8- and 12-channel pipetters
  • Autoclavable
  • Non-sterile version includes lid
Specifications
Specifications
Transparent
Polypropylene
Reagent Reservoir
Individually Wrapped
Autoclavable
Limited-use
60 mL
Sterile
For use with multichannel pipets. Without lid.
100/Pk.
1
V-bottom
Product Suggestions
Product Suggestions