missing translation for 'onlineSavingsMsg'
Learn More

BRANDTECH™ BRAND™ Bottletop Dispenser Replacement Discharge Tube

Replacement discharge tubes for use with seripettor™ / seripettor™ pro bottle-top dispensers.

Supplier:  BRANDTECH™ BRAND™ 707916

 View more versions of this product

Catalog No. 1387984


Only null left

Description

Description

Discharge tubes for seripettor™. Includes closure cap, EPDM discharge valve, and sealing ring:

  • Part No: 704520 - standard tip, for use with 10 and 25 mL models

Discharge tube for seripettor™ pro. Includes integrated discharge valve and flat seal.

  • Part No: 707915 - fine tip, for use with 2 mL model
  • Part No. 707916 - standard tip, for use with 10 mL model
  • Part No. 707918 - standard tip, for use with 25 mL model

Discharge tube adapter sold separately

Discharge tube for seripettor™ pro. Includes a Luer-Lock attachment for microfilter

  • Part No. 707928 - for use with 2, 10, and 25 mL models.

Discharge tube adapter sold separately. Not suitable for HF and peroxide.

Specifications

Specifications

Discharge Tube
Red
BRAND seripettor pro bottle-top dispenser
Standard
90 mm
Autoclavable
Serial dispensing
Integrated valve
3.54 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions