missing translation for 'onlineSavingsMsg'
Learn More

BRANDTECH™ VITLAB™ Reagent Bottles with Screw Caps

Catalog No. 14379351
Change view
Click to view available options
Capacity (Metric):
1000 mL
5000 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
14379351 1000 mL
361020449 5000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14379351 Supplier BRANDTECH™ VITLAB™ Supplier No. 100689

VITLAB™ polypropylene reagent bottles with polypropylene screw caps have good chemical resistance. Autoclavable at 121 C for 20 minutes.

  • Good chemical resistance for long-term storage of liquids.
  • Screw caps help protect against outside contamination and leakage.
  • Autoclavable at 121 C, according to DIN EN 285

Specifications

Capacity 33.8 oz., 1000mL
Capacity (Metric) 1000 mL
Capacity (English) 33.8 oz.
Color Transparent
Diameter (English) 4.25 in.
Material Polypropylene
Quantity 10/Pk.
Sterility Non-sterile
Closure Included Yes
Graduated No
Mouth Threaded
Height (Metric) 202 mm
Product Type Reagent Bottle
Height (English) 7.95 in.
Closure Material Polypropylene
Closure Size GL 32
Diameter (Metric) 108 mm
Closure Type Screw Cap
Autoclavable Autoclavable
Show More Show Less