Learn More
BrandTech™ BRAND™ BRANDplates cellGrade™ 96-well PS Microplates
Sterile 96-well microplates with cellGrade™surface treatment for optimal growth. Ideal for cultivating adherent cells with a hydrophilic PS surface featuring accessible carboxyl and hydroxyl groups. Enhances serum component binding for effective indirect cell adhesion.
$876.37 - $2467.26
Specifications
Barcode No Barcode
Certifications/Compliance ISO 11137, DIN EN ISO 10993
Format Standard
Lid With Lid
Array 8 x 12
View More Specs
Products 6
Catalog Number Mfr. No. Color Well Shape Price Quantity  
Catalog Number Mfr. No. Color Well Shape Price Quantity  
13-882-251
View Documents
N/A Black, Clear Bottom, Clear Bottom Flat Bottom (F-well)
Pack of 50 Each for $2,467.26
Only null left
13-882-244
View Documents
N/A Clear Round Bottom (U-well)
Pack of 50 Each for $876.37
Only null left
13-882-246
View Documents
N/A Clear Flat Bottom (F-well)
Pack of 50 Each for $876.37
Only null left
13-882-248
View Documents
N/A White Flat Bottom (F-well)
Pack of 50 Each for $890.74
Only null left
13-882-249
View Documents
N/A Black Flat Bottom (F-well)
Pack of 50 Each for $876.37
Only null left
13-882-250
View Documents
N/A White, Clear Bottom, Clear Bottom Flat Bottom (F-well)
Pack of 50 Each for $2,467.26
Only null left
Description
Description
  • Hydrophilic surface with freely-accessible carboxyl and hydroxyl groups
  • Easily binds serum components to for indirect adhesion of cells
  • Sterilized by B-radiation to ISO 11137 and AAMI guidelines for a SAL of 10-6
  • Meet the requirements of Ph. Eur. and the USP 29
  • BIO-CERT™ CELL CULTURE QUALITY means inserts: free of pyrogens, endotoxins, human DNA, RNase, and DNase
  • Endotoxins are <0.01 EU/mL
  • Orange alphanumeric embossing for easy well identification
Order Info
Packs of 50 individually wrapped plates with lids
Specifications
Specifications
No Barcode
Standard
8 x 12
Polystyrene
Sterile
Cultivation of adherent cell lines
ISO 11137, DIN EN ISO 10993
With Lid
96-well microplate
96
cellGrade
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'