Learn More
BrandTech™ BRAND™ BRANDplates cellGrade™ 96-well PS Microplates
Sterile 96-well microplates with cellGrade™surface treatment for optimal growth. Ideal for cultivating adherent cells with a hydrophilic PS surface featuring accessible carboxyl and hydroxyl groups. Enhances serum component binding for effective indirect cell adhesion.
 View more versions of this product
Catalog No. 13-882-250
Only null left
Explore available promotions
Description
Description
  • Hydrophilic surface with freely-accessible carboxyl and hydroxyl groups
  • Easily binds serum components to for indirect adhesion of cells
  • Sterilized by B-radiation to ISO 11137 and AAMI guidelines for a SAL of 10-6
  • Meet the requirements of Ph. Eur. and the USP 29
  • BIO-CERT™ CELL CULTURE QUALITY means inserts: free of pyrogens, endotoxins, human DNA, RNase, and DNase
  • Endotoxins are <0.01 EU/mL
  • Orange alphanumeric embossing for easy well identification
Order Info
Packs of 50 individually wrapped plates with lids
Specifications
Specifications
No Barcode
Standard
8 x 12
White, Clear Bottom, Clear Bottom
Polystyrene
96
cellGrade
Flat Bottom (F-well)
50/Cs.
ISO 11137, DIN EN ISO 10993
With Lid
Carboxyl and Hydroxyl Groups
96-well microplate
330 μL
Sterile
330 μL
Cultivation of adherent cell lines
1/Pk.
Product Suggestions
Product Suggestions