Learn More
BrandTech™ BRAND™ BRANDplates cellGrade™ plus, 96-well Flat Bottom Microplates, PS
Sterile 96-well microplates with cellGrade™plus surface treatment for optimal cell growth. Features protein-like surface for fast cell attachment and better yield. Ideal for cultivating sensitive or fastidious cell lines, even in reduced-serum media
 View more versions of this product
Catalog No. 13-882-255
Only null left
Explore available promotions
Description
Description
  • Hydrophilic surface with freely-accessible amino, carboxyl and hydroxyl groups
  • Protein-like composition allows for direct cellular attachment
  • Sterilized by B-radiation to ISO 11137 and AAMI guidelines for a SAL of 10-6
  • Meet the requirements of Ph. Eur. and the USP 29
  • BIO-CERT™ CELL CULTURE QUALITY: free of pyrogens, endotoxins, human DNA, RNase, and DNase
  • Endotoxins are <0.01 EU/mL
  • Orange alphanumeric embossing for easy well identification
Order Info
Packs of 50 individually wrapped plates with lids
Specifications
Specifications
No Barcode
ISO 11137, DIN EN ISO 10993
With Lid
1/Pk., 50/Cs.
96-well microplate
330 μL
Sterile
330 μL
Cultivation of cells in reduced serum media and fastidious cell lines
1/Pk.
Hydrophilic
Standard
8 x 12
White, Clear Bottom, Clear Bottom
Polystyrene
96
cellGrade Plus
Flat Bottom (F-well)
50/Cs.
Clear
Product Suggestions
Product Suggestions