missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BrandTech™ BRAND™ BRANDplates cellGrade™ plus, 96-well Flat Bottom Microplates, PS
Sterile 96-well microplates with cellGrade™plus surface treatment for optimal cell growth. Features protein-like surface for fast cell attachment and better yield. Ideal for cultivating sensitive or fastidious cell lines, even in reduced-serum media
Description
- Hydrophilic surface with freely-accessible amino, carboxyl and hydroxyl groups
- Protein-like composition allows for direct cellular attachment
- Sterilized by B-radiation to ISO 11137 and AAMI guidelines for a SAL of 10-6
- Meet the requirements of Ph. Eur. and the USP 29
- BIO-CERT™ CELL CULTURE QUALITY: free of pyrogens, endotoxins, human DNA, RNase, and DNase
- Endotoxins are <0.01 EU/mL
- Orange alphanumeric embossing for easy well identification
Order Info
Packs of 50 individually wrapped plates with lids
Specifications
| No Barcode | |
| ISO 11137, DIN EN ISO 10993 | |
| With Lid | |
| Clear | |
| Polystyrene | |
| 96 | |
| cellGrade Plus | |
| Flat Bottom (F-well) | |
| 50/Cs. | |
| Clear |
| Hydrophilic | |
| Standard | |
| 8 x 12 | |
| 96-well microplate | |
| 350 μL | |
| Sterile | |
| 350 μL | |
| Cultivation of cells in reduced serum media and fastidious cell lines | |
| 1/Pk. |