missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Warning: Authorized Personnel Only Signs

Identify areas designated for certain personnel and direct facility traffic

Supplier:  Brady™ 69566

 View more versions of this product

Catalog No. 19-035647

Only null left
Explore available promotions

Description

Description

  • Available in plastic, self sticking polyester and premium fiberglass materials
  • Black on orange
  • Language: english
  • Available sizes (W x H): 10 x 7 in. or 14 x 10 in.

Specifications

Specifications

ANSI Z535, OSHA 1910.145, ASTM D 1593, ASTM D 638, ASTM D 790, ASTM D 646
1 Ea.
Fiberglass Reinforced Polyester
Use in industrial, utility, commercial and institutional environments, common uses include safety signs, utility signs, information signs, traffic signs and valve tags, the sign construction is recommended for both indoor and outdoor use
25.4 cm
-40°F
14 in.
-40°C to 90°C
Warning Authorized Personnel Only
Black on Orange
Fiberglass, Polyester
9000 psi
Surface Mount
10 in.
193°F
0.1 in.
35.5 cm
-40°F to 193°F
Admittance Sign
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions