missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Universal Ball Valve Lockout Devices

Catalog No. 189991332
Change view
Click to view available options
Product Type:
Large Ball Valve Lockout
Small Blocking Arm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
189991332 Small Blocking Arm
189991334 Large Ball Valve Lockout
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 189991332 Supplier Brady™ Supplier No. 65402

Ideal for valves on insulated pipe

  • Designed to lock out almost any quarter-turn valve regardless of valve shape—accommodates round, square, and flat valves
  • Can be used in any position (even upside down) to work with odd-shaped, straight, or angled valve handles
  • For nonlockout applications, device can lock valves in the ON and throttled positions
  • Accepts additional arm (available separately) to lock handles in throttled position and lock out three-, four-, or five-way valves
  • Devices and arms made of durable nylon and steel
  • Lightweight and resistant to most chemicals

Order Info

Valve fits handles up to 1.25 in. thick (3.2cm) and 1.63 in. W (4.1cm).

Specifications

Product Type Small Blocking Arm
Color Red
Material Nylon
For Use With (Application) To lock out almost any quarter-turn valve regardless of valve shape
For Use With (Equipment) Small universal valve lockout
Temperature Range (English) −20° to +120°F
Temperature Range (Metric) 0° to 250°C