Learn More
Brady™ ToughStripe™ Floor Marking Tape
ToughStripe™ Floor Marking Tape (3.5 m/100 ft. in length) is a long-lasting, heavy-duty solution for marking floors, aisles, work cells and other designated areas.
Supplier:  Brady™ 104371
 View more versions of this product
Catalog No. 19150721
Only null left
Description
Description
  • Visible floor-marking improves safety and organization in almost any industrial environment
  • High-gloss surface shines like new paint and withstands forklift traffic without tearing or lifting
  • Ideal for marking aisles, passageways and storage locations and identifying equipment and egress paths
  • Can be easily applied by one person working alone
Specifications
Chemical Identifiers
None
Tape
None
Used to mark aisles, passageways, storage locations and point out safety equipment and exits
1 Roll/Pack
Rectangle
0.008 in.
30.48 m
Gloss
Polyester
Non-sterile
-18°C
1 Roll
-18°C to +54°C
100 ft.
Specifications
ASTM ANSI F1679, ANSI A1264.2 2006, OSHA 1910.22
Rubber, Permanent
White
Tape
Single-side Printable
Label Tape
Used to mark aisles, passageways, storage locations and point out safety equipment and exits
-18°C
1 Roll
0°F to 130°F
0.008 in.
100 ft. x 4 in. (30.48 m x 10.2 cm)
30.48 m
10.2 cm
ToughStripe FLOOR TAPES
None
Gloss
Polyester
None
Non-sterile
115 Gardner Units
1 Roll/Pack
Rectangle
-18°C to +54°C
0.201 mm
100 ft.
4 in.
TS-101.60-514-WT-RL
Product Suggestions
Product Suggestions