Learn More
Brady™ ToughStripe™ Floor Marking Center Marks
ToughStripe™ Floor Marking Center Marks are extremely durable die-cut floor marking shapes made of rugged polyester with ultra-aggressive adhesive and hold up against forklift traffic.
Supplier:  Brady™ 104476
 View more versions of this product
Catalog No. 19150806
Only null left
Description
Description
  • Visible floor-marking improves safety and organization in almost any industrial environment
  • High-gloss surface shines like new paint and withstands forklift traffic without tearing or lifting
  • Ideal for marking corners of work cells and storage locations
  • Can be easily applied by one person working alone
Specifications
Chemical Identifiers
ASTM ANSI F1679, ANSI A1264.2 2006, OSHA 1910.22
Rubber, Permanent
Gloss
Polyester
None
Non-sterile
115 Gardner Units
20 Pieces/Pack
Cross
-18°C to +54°C
0.201 mm
ToughStripe Diecut Shapes (L's, T's, +'s, footprints and more)
None
Tape
Single-side Printable
Label Tape
For floor corner mark, Used to mark aisles, passageways, storage locations and point out safety equipment and exits
-18°C
20/Pk.
0°F to 130°F
0.008 in.
Specifications
ASTM ANSI F1679, ANSI A1264.2 2006, OSHA 1910.22
Rubber, Permanent
Black
Tape
Single-side Printable
Label Tape
For floor corner mark, Used to mark aisles, passageways, storage locations and point out safety equipment and exits
-18°C
20/Pk.
0°F to 130°F
0.008 in.
203.20 x 76.2 mm
20.32 cm
76.2 mm
203.20 mm
ToughStripe Diecut Shapes (L's, T's, +'s, footprints and more)
None
Gloss
Polyester
None
Non-sterile
115 Gardner Units
20 Pieces/Pack
Cross
-18°C to +54°C
0.201 mm
8 in.
3 in.
8 in.
TS-76.20-514-CEN-BK-20
Product Suggestions
Product Suggestions