Learn More
Brady™ ToughStripe™ Floor Arrows
ToughStripe™ Floor Arrows are extremely durable die-cut floor marking shapes made of rugged polyester with ultra-aggressive adhesive that holds up against forklift traffic.
Supplier:  Brady™ 104414
 View more versions of this product
Catalog No. 19150744
Only null left
Description
Description
  • Visible floor-marking improves safety and organization in almost any industrial environment
  • High-gloss surface shines like new paint and withstands forklift traffic without tearing or lifting
  • Ideal for marking aisles, passageways and storage locations and identifying equipment and egress paths
  • Can be easily applied by one person working alone
Specifications
Chemical Identifiers
ANSI A1264.2-2006, OSHA 1910.22
Polyester
ToughStripe Diecut Shapes (L's, T's, +'s, footprints and more)
129°F
2 mil
Arrow
Arrow
High Gloss
100/Pk.
Ideal for marking aisles, passageways and storage locations and pointing out important safety equipment and egress paths
0°F
-18°C to 54°C
0°F to 129 °F
Floor Arrows
Specifications
ANSI A1264.2-2006, OSHA 1910.22
High Gloss
100/Pk.
ToughStripe Diecut Shapes (L's, T's, +'s, footprints and more)
5 in.
129°F
2 mil
50.8 mm
Arrow
50.80 x 127 mm
Floor Arrows
TS-50.80X127.00-514-ARO-BL-100
Polyester
Permanent Rubber
Ideal for marking aisles, passageways and storage locations and pointing out important safety equipment and egress paths
127 mm
0°F
2 in.
-18°C to 54°C
0°F to 129 °F
Arrow
Blue
Product Suggestions
Product Suggestions