missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Spill Response Plus Universal Absorbents
Description
Spécifications
Spécifications
| Color | Gray |
| Capacity (English) | 3.3 gal. |
| Material | Polypropylene |
| Type | Universal Sorbent |
| For Use With (Application) | Water-based liquids and oils |
| Packaging Quantity | 1 Ea. |
| Weight Capacity | Heavyweight |