missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Spill Response Plus Universal Absorbents

Catalog No. 19140736
Change view
Click to view available options
Capacity (English):
3.3 gal.
5.5 gal.
11.1 gal.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
19140736 5.5 gal.
19140735 11.1 gal.
19140741 3.3 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19140736 Supplier Brady™ Supplier No. SRP100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Absorbs just about any liquid

  • Small convenient size for employees to fold and carry in pockets for quick response time
  • Highly absorbent, multi-purpose absorbent to deal with water-based liqiuds and oils
  • Dimpled for added durability and reduced linting
  • Available as pads and rolls
  • Specifications

    Color Gray
    Capacity (English) 5.5 gal.
    Material Polypropylene
    Type Universal Sorbent
    For Use With (Application) Water-based liquids and oils
    Packaging Quantity 100/Cs.
    Weight Capacity Heavyweight