Learn More
Brady™ PowerMark™ Polyester Tapes
PowerMark™ Polyester Tapes is Hi-Performance tape (B-569) with a low halide content that is designed for marking stainless steel pipes particularly in harsh environments.
Supplier:  Brady™ 13575
 View more versions of this product
Catalog No. 19-041-090
Only null left
Explore available promotions
Description
Description
  • Durable, ultra-smooth polyester labels with permanent pressure sensitive acrylic adhesive
  • Outdoor grade polyester tape can be thermal transfer printed
  • Withstands mild chemicals, oil and water
  • Service temperature of -40°F to 212°F (-40°C to 100°C)
Specifications
Specifications
Acrylic, Pressure-sensitive
Gloss
Polyester
Thermal Transfer
Non-sterile
BBP™85 Industrial Sign and Label Printer, PowerMark™ Industrial Sign and Label Printer
3 to 5 Years
-40°F to 212°F
0.003 in.
50 ft.
7 in.
Orange
Tape
Single-side Printable
Label Tape
Ideal for mounting on sign panels
80 Gardner Units
1 Ea.
-40°C to +100°C
50 ft. x 7 in.
15.24 m
Product Suggestions
Product Suggestions