missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Oversized Breaker Lockout

Catalog No. 19807177
Change view
Click to view available options
Color:
Gray
Red
Material:
Galvanized Steel
Nylon
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Material
19807177 Gray Galvanized Steel
19055386 Red Nylon
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19807177 Supplier Brady™ Supplier No. 65319

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Oversized Breaker Lockout cables provide solutions for locking out gate valves, handles and other oversized devices.

  • Brady's Original Cable Lockout Device is constructed of impact modified nylon
  • Cable for use with Original Cable Lockout.

Specifications

Product Type Oversized Breaker Lockout
Color Gray
Material Galvanized Steel
Certifications/Compliance OSHA 1910.147
Diameter (English) 0.125 in.
Diameter (Metric) 0.3 cm
For Use With (Equipment) Fits breakers up to 2.25 W x 0.87 in. thick
Length (English) 8 ft.
Length (Metric) 2.4 m