Learn More
Brady™ Overlaminate Tape
Overlaminate Tape is applied to labels for added protection and extended durability.
Supplier:  Brady™ 25966
 View more versions of this product
Catalog No. 19806978
Only null left
Description
Description
  • The B-674 material from Brady™ is a clear 1.0 mil polyester film with an acrylic pressure sensitive adhesive
  • Service temperature range of -40 to 194° F
  • Average outdoor durability up to 2 years
Specifications
Chemical Identifiers
Acrylic, Pressure-sensitive
Polyester
Used as a clear over laminate
100 Gardner Units
1 Roll
-40°C to +90°C
0.0019 in.
9.14 m
Clear
Non-sterile
GlobalMark™ Industrial Label Maker
Up to 2 Years
-40 to 194°F
21 lb./in.
30 ft.
Specifications
Acrylic, Pressure-sensitive
Polyester
Used as a clear over laminate
100 Gardner Units
1 Roll
-40°C to +90°C
0.0019 in.
9.14 m
Clear
Non-sterile
GlobalMark™ Industrial Label Maker
Up to 2 Years
-40 to 194°F
21 lb./in.
30 ft.
0.75 in.
Product Suggestions
Product Suggestions