Learn More
Brady™ Nylon Cloth Laboratory Labels
Nylon Cloth Laboratory Labels (B-499) wrap-around labels are highly pliable to conform to tightly curved surfaces such as wire and cable.
Specifications
Autoclavable Autoclavable
Certifications/Compliance RoHS
Adhesive Type Acrylic, Permanent
Color White
Display Consignment
View More Specs
Products 2
Catalog Number Mfr. No. Width (English) Height (English) Height (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Width (English) Height (English) Height (Metric) Price Quantity & Availability  
19140656
View Documents Promotion Details
Brady™
B33152499
1 in. 0.3 in. 9 mm N/A
Only null left
19140687
View Documents Promotion Details
Brady™
B33151499
2 in. 0.25 in. 6 mm N/A
Only null left
Description
Description
  • Matte white polyamide coated nylon cloth material has a permanent acrylic adhesive and is thermal transfer printable
  • Recommended for indoor use
  • RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC; UL approved; CSA accepted
  • Service temperature of -40 to 193° F (-40 to 90° C)
Specifications
Specifications
Autoclavable
Acrylic, Permanent
Consignment
Die-cut
Single-side Printable
Laboratory Label
Wiremarking and general labeling applications
-40°C
1500/Pk.
-40°F to 193°F
80 lb./in.
RoHS
White
Matte
Nylon Cloth
Thermal Transfer
Non-sterile
BBP™ 33
1500 Labels/Box
Rectangle
-40°C to +90°C
0.0065 in.