Learn More
Brady™ No Smoking Signs
No Smoking safety signs from Brady™ help to manage and sustain a clean and safe workplace and are available in a variety of materials and sizes that best fit your application.
Supplier:  Brady™ 47203
 View more versions of this product
Catalog No. 19-807-070
Only null left
Explore available promotions
Description
Description
  • Self-Sticking Polyester (B-302) is an Indoor/Outdoor high performance pressure sensitive sign that is surface printed polyester with a polyester overlaminate.
  • Plastic signs (B-401) are ideal for use in light duty industrial, utility, commercial and institutional environments.
  • Extremely durable (B-120) Premium Fiberglass will not chip, fade, rust, shatter or peel for up to 25 years.
  • Durable Non-Reflective Aluminum (B-555) is a rigid aluminum panel with protected graphics designed for use in industrial, utility, commercial and institutional environments.
Specifications
Specifications
ASTM, OSHA
Fiberglass, Polyester
9000 psi
Fiberglass Reinforced Polyester
10 in.
-40°F
14 in.
-40°C to 90°C
10 x 14 in.
No Smoking Sign
Industrial
1 Ea.
Non-adhesive
Indoor and outdoor use, industrial, utility, commercial and institutional environment
193°F
0.1 in.
35.56 cm
-40°F to 193°F
DANGER No Smoking, Matches or Open Lights
Red on White
Product Suggestions
Product Suggestions