Learn More
Brady™ No Smoking Signs
No Smoking safety signs from Brady™ help to manage and sustain a clean and safe workplace and are available in a variety of materials and sizes that best fit your application.
Supplier:  Brady™ 42662
 View more versions of this product
Catalog No. 19-807-046
Only null left
Explore available promotions
Description
Description
  • Self-Sticking Polyester (B-302) is an Indoor/Outdoor high performance pressure sensitive sign that is surface printed polyester with a polyester overlaminate.
  • Plastic signs (B-401) are ideal for use in light duty industrial, utility, commercial and institutional environments.
  • Extremely durable (B-120) Premium Fiberglass will not chip, fade, rust, shatter or peel for up to 25 years.
  • Durable Non-Reflective Aluminum (B-555) is a rigid aluminum panel with protected graphics designed for use in industrial, utility, commercial and institutional environments.
Specifications
Specifications
ASTM, OSHA
1 Ea.
Indoor and outdoor use, industrial, utility, commercial and institutional environment
230°F
0.04 in.
35.56 cm
-40°F to 230°F
DANGER No Smoking or Open Light Within-feet
Red on White
Aluminum
Non-adhesive
10 in.
-40°F
14 in.
-40°C to 110°C
10 x 14 in.
No Smoking Sign
Product Suggestions
Product Suggestions