Learn More
Brady™ NFPA Placard Systems
Catalog No. 19807076
Change view
Click to view available options
Dimensions (L x W):
10 x 10 in.
11 x 11 in.
Material:
Fiberglass
Vinyl
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W) Material
19807076 11 x 11 in. Fiberglass
19807075 10 x 10 in. Vinyl
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19807076 Supplier Brady™ Supplier No. 50055
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Customize placards on the spot with NFPA squares blue, white, red and yellow
  • Available in fiberglass or vinyl
  • Includes 10 blank placards and 10 characters each of numbers 0-4, symbols for Use No Water, Radiation, OXY, ACID, COR, and ALK
Specifications
Material Fiberglass
Shape Diamond
Languages English
Dimensions (L x W) 11 x 11 in.
Product Type NFPA Placard Kit
Color Yellow