Learn More
Brady™ MRO Plus™ Universal Dispense-N-Dispose Heavy Pads
Catalog No. 296353201
Change view
Click to view available options
Capacity (English):
39 gal./Bale
Catalog No. Capacity (English)
296353201 39 gal./Bale
1 options
Catalog No. 296353201 Supplier Brady™ Supplier No. MRO150DND
Absorbs all industrial liquids (water, petroleum-based and chemical-based fluids)
  • Three-ply construction
  • Perforated - to reduce waste
  • Gray color masks drips and spills
  • Includes the MRO100 pads in Dispense-N-Dispose system
Specifications
Color Gray
Capacity (English) 39 gal./Bale
Material Polypropylene
Type Universal Dispense-N-Dispose Heavy Pad
For Use With (Application) Overspray leaks and drips, workstations, heavy clean-up and wipe-up, drums - liquid storage, outfalls and discharge ponds