Learn More
Brady™ Machine and Operational Signs
Machine and Operational Signs are made of durable non-reflective aluminum (B-555). The rigid aluminum panel with protected graphics is designed for use in industrial, utility, commercial and institutional environments.
Supplier:  Brady™ 42543
 View more versions of this product
Catalog No. 19807041
Only null left
Description
Description
  • Non-Reflective Aluminum (B-555) is ideal for indoor and outdoor usage
  • Printed with durable inks and clear-coat protection for fade resistance
  • Withstands wind, rain, sunlight and high temperatures
  • Average outdoor durability 5-8 years
Specifications
Chemical Identifiers
ANSI Z535, OSHA 1910.145
Aluminum
Non-adhesive
Use in industrial, utility, commercial and institutional environments
-40°F
-40°C to 110°C
Machine and Operational Sign
Industrial
1 Ea.
Surface Mount
230°F
0.03 in.
-40°F to 230°F
Specifications
ANSI Z535, OSHA 1910.145
Aluminum
Non-adhesive
Use in industrial, utility, commercial and institutional environments
230°F
0.03 in.
35.56 cm
-40°F to 230°F
DANGER Moving Machinery
Red on White
Industrial
1 Ea.
Surface Mount
10 in.
-40°F
14 in.
-40°C to 110°C
10 x 14 in.
Machine and Operational Sign
Product Suggestions
Product Suggestions