Learn More
Brady™ High Visibility Signs - Rigid
Rigid High Visibility Signs from Brady™ feature bright colors, graphics and pictograms to effectively communicate a safety message or identify the location of a given item.
Supplier:  Brady™ 45373
 View more versions of this product
Catalog No. 19-054-827
Only null left
Explore available promotions
Description
Description
  • V-style 45° high visibility signs project from the wall to be easily viewed from all angles
  • Designed to be seen from all access points in your facility
  • Pre-drilled holes for easy installation
  • Available in rigid polyethylene (B-450) or rigid polystrene (B-493)
Specifications
Specifications
ANSI Z535, OSHA 1910.145
Polystyrene
Non-adhesive
194°F
0.1 in.
26.416 cm
-40°F to 194°F
Women
White on Blue
Industrial
1 Ea.
5.5 in.
-40°F
10 in.
-40°C to 90°C
5.5 x 10.4 in.
High Visibility Sign
Product Suggestions
Product Suggestions