missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Hazardous Information Placards

Catalog No. 19016336
Change view
Click to view available options
Displayed Text:
Color Bar Version
Combined Version- NFPA Plus Rating Explanation Guide
Height (English):
10 in.
14 in.
Width (English):
10 in.
14 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Displayed Text Height (English) Width (English)
19016336 Color Bar Version 10 in. 14 in.
19807256 Combined Version- NFPA Plus Rating Explanation Guide 14 in. 10 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19016336 Supplier Brady™ Supplier No. 58727
Only null left

Hazardous Information Placards are made of extremely durable B-120 Premium Fiberglass that will not chip, fade, rust, shatter or peel for up to 25 years.

  • Graphics are permanently embedded within the sign panel, allowing for use in the most severe environments
  • Excellent chemical and abrasion resistance
  • Ideal for both indoor and outdoor applications
  • Service temperature of -40°F to 193°F (-40°C to 90°C)

Specifications

Certifications/Compliance Globally Harmonized System of Classification and Labeling of Chemicals (GHS); NFPA 704; OSHA 1910.1200
Includes NFPA version or combined version , four mounting hole for easy installation
Material Fiberglass, Polyester
Quantity 1 Ea.
Tensile Strength 9000 psi
Adhesive Type Non-adhesive
Description Fiberglass Reinforced Polyester
Mounting Type Surface Mount
For Use With (Application) Designed for use in industrial, utility, commercial and institutional environments
Height (English) 10 in.
Max. Temperature (English) 193°F
Min. Temperature (English) -40°F
Thickness 0.1 in.
Width (English) 14 in.
Width (Metric) 35.56 cm
Temperature Range (Metric) -40°C to 90°C
Temperature Range (English) -40°F to 193°F
Dimensions (L x W) 10 x 14 in.
Displayed Text Color Bar Version
Product Type Hazardous Information Placard
Color Red
Show More Show Less