missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Globally Harmonized System (GHS) Gas Pressure Picto Labels
For gases under pressure
$10.58 - $13.88
Specifications
| Certifications/Compliance | GHS, OSHA 29 CFR 1910.1200 |
|---|---|
| Color | Black, Red, White |
| Format | Label |
| Sterility | Non-sterile |
| For Use With (Application) | For gasses under pressure |
| Catalog Number | Mfr. No. | Quantity | Material | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Material | Price | Quantity & Availability | ||||
|
19140515
|
Brady™
118833 |
39/Pk. | Vinyl |
Pack of 39 Each for $13.88
|
|
||||
|
19140524
|
Brady™
118887 |
39/Pk. | Paper |
Pack of 39 Each for $10.93
|
|
||||
|
19140533
|
Brady™
118834 |
16/Pk. | Vinyl |
Pack of 16 Each for $12.26
|
|
||||
|
19140542
|
Brady™
118888 |
16/Pk. | Paper |
Pack of 16 Each for $10.58
|
|
||||
|
19140702
|
Brady™
118835 |
4/Card | Vinyl (B-7569) |
Pack of 4 Each for $11.89
|
|
||||
Description
Pre-printed die cut labels identify hazardous products for chemical risks, health risks and environmental risks in accordance to the United Nations' universal approach to classifying and communicating chemical hazards
- Legend: Gas Pressure
- Made of B-7569 Viny or B-235 Paper
- Available in three sizes
Specifications
| GHS, OSHA 29 CFR 1910.1200 | |
| Label | |
| For gasses under pressure |
| Black, Red, White | |
| Non-sterile |