Learn More
Brady™ Globally Harmonized System (GHS) Gas Pressure Picto Labels
For gases under pressure
Supplier:  Brady™ 118833
 View more versions of this product
Catalog No. 19140515
Only null left
Description
Description
Pre-printed die cut labels identify hazardous products for chemical risks, health risks and environmental risks in accordance to the United Nations' universal approach to classifying and communicating chemical hazards
  • Legend: Gas Pressure
  • Made of B-7569 Viny or B-235 Paper
  • Available in three sizes
Specifications
Chemical Identifiers
GHS, OSHA 29 CFR 1910.1200
Label
For gasses under pressure
Black, Red, White
Non-sterile
Specifications
GHS, OSHA 29 CFR 1910.1200
Black, Red, White
Label
Pictogram Label
GHS Gas Pressure
3 Years
-4°F to 167°F
0.004 in.
0.6 in.
15.20 mm
Acrylic, Permanent
Gloss
Vinyl
Non-sterile
For gasses under pressure
39/Pk.
-20°C to +75°C
0.625 x 0.625 in.
0.62 in.
Product Suggestions
Product Suggestions