missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Danger: Wear Face Shield And Rubber Gloves... Signs
Identify equipment, rooms and materials that contain or are contaminated with hazardous agents.
Supplier: Brady™ 70582
Description
- Bright colors and bold text provide high visibility to critical messages
- Sign reads “DANGER - WEAR FACE SHIELD AND RUBBER GLOVES WHEN HANDLING CHEMICALS”
- Black/red on white
- Available in different sizes and materials
Chemical Identifiers
| 1 Ea. | |
| -40°F | |
| Protective Wear Sign |
| Surface Mount | |
| DANGER Wear Face Shield And Rubber Gloves When Handling Chemicals | |
| Red on White |
Specifications
| ASTM D 1593, D 638, D 790, D 646, D 635 | |
| 1 Ea. | |
| Premium Fiberglass | |
| Industrial, utility, commercial and institutional environments for both indoor and outdoor use | |
| 25.4 cm | |
| -40°F | |
| 14 in. | |
| -40°C to 90°C | |
| DANGER Wear Face Shield And Rubber Gloves When Handling Chemicals | |
| Red on White |
| Fiberglass | |
| 9000 psi | |
| Surface Mount | |
| 10 in. | |
| 193°F | |
| 0.1 in. | |
| 35.5 cm | |
| -40°F to 193°F | |
| Protective Wear Sign |