Learn More
Brady™ Compact Keyed-Different 1 in. Shackle Aluminum Padlocks
Compact Keyed-Different 1 in. Shackle Aluminum Padlocks include two keys per lock.
Supplier:  Brady™ 133266
 View more versions of this product
Catalog No. 19103520
Only null left
Description
Description
  • Compact lock body measures 1-1/4 in. H x 1-1/4 in. W
  • Bright, anodized aluminum finish provides greater visibility and fade-resistance in all environoments
  • Reserved, paracentric keyway provides optimal security (prevents someone from getting a copy made at a local key shop)
  • Each padlock is keyed differently with keys precision-machined to exacting standards, so that the key for one lock will not open another in the set
Specifications
Chemical Identifiers
OSHA 1910.147
Aluminum
Outdoor; Industrial Manufacturing; Food and Beverage; Confined Spaces
1 in.
Keyed Different
Padlock
Outdoor; Industrial Manufacturing; Food and Beverage; Confined Spaces
Specifications
OSHA 1910.147
Keyed Different
6/Pk.
Outdoor; Industrial Manufacturing; Food and Beverage; Confined Spaces
1 in.
Black
Aluminum
Padlock
Outdoor; Industrial Manufacturing; Food and Beverage; Confined Spaces
Product Suggestions
Product Suggestions