missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Chemical and Hazardous Materials Signs: DANGER - FLAMMABLE MATERIALS

Catalog No. 1704622
Change view
Click to view available options
Material:
Aluminum
Fiberglass
Plastic
Height (Metric):
25.4 cm
25.40 cm
Width (Metric):
35.5 cm
35.56 cm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Height (Metric) Width (Metric)
1704622 Fiberglass 25.40 cm 35.56 cm
19035964 Plastic 25.40 cm 35.56 cm
19000659 Aluminum 25.4 cm 35.5 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 1704622 Supplier Brady™ Supplier No. 72246
Only null left

Helps identify plant's chemical storage and hazardous materials areas

  • Available in three types of materials: fiberglass, self-sticking polyester and plastic
  • Available in various widths and heights
  • Black/red on white

Specifications

Certifications/Compliance ISO 9001, ANSI Z535, OSHA 1910.145
Material Fiberglass
Quantity 1 Ea.
Tensile Strength 9000 psi
Mounting Type Surface Mount
For Use With (Application) Designed for use in industrial, utility, commercial and institutional environments
Height (English) 10 in.
Height (Metric) 25.40 cm
Max. Temperature (English) 193°F
Min. Temperature (English) -40°F
Thickness 0.1 in.
Width (English) 14 in.
Width (Metric) 35.56 cm
Temperature Range (Metric) -40°C to 90°C
Temperature Range (English) -40°F to 193°F
Displayed Text Flammable Material
Product Type Hazardous Materials Sign
Color Red on White
Show More Show Less