missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Chemical and Hazardous Materials Signs: DANGER - FLAMMABLE GAS
Identify equipment, rooms and materials which contain or are contaminated with hazardous agents
$20.40 - $46.46
Specifications
| Quantity | 1 Ea. |
|---|---|
| Mounting Type | Surface Mount |
| For Use With (Application) | Designed for use in industrial, utility, commercial and institutional environments |
| Height (English) | 10 in. |
| Height (Metric) | 25.40 cm |
Description
- Available in three materials - plastic, fiberglass and polyester
- Available in various widths and heights
- Black/red on white
Specifications
| 1 Ea. | |
| Designed for use in industrial, utility, commercial and institutional environments | |
| 25.40 cm | |
| 14 in. | |
| Flammable Gas | |
| Red on White |
| Surface Mount | |
| 10 in. | |
| -40°F | |
| 35.56 cm | |
| Hazardous Materials Sign |