Learn More
Brady™ Caution and Warning Tapes
Various tape designs in choice of two materials
$18.62 - $126.62
| Catalog Number | Mfr. No. | Color | Dimensions (L x W) | dimMaterial | Description | Price | Quantity & Availability | ||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Color | Dimensions (L x W) | dimMaterial | Description | Price | Quantity & Availability | ||
|
19016186
|
Brady™
55294 |
Red on White | 54 ft. x 1 in. | Stripes |
Each for $35.84
|
|
|||
|
19016190
|
Brady™
BRD55302 |
Black on Yellow | 54 ft. x 2 in. | Stripes |
Each for $76.55
|
|
|||
|
19016185
|
Brady™
55292 |
Black on White | 54 ft. x 2 in. | Stripes |
Each for $76.55
|
|
|||
|
19016191
|
Brady™
55303 |
Black on Yellow | 54 ft. x 3 in. | Stripes |
Each for $99.10
|
|
|||
|
19017131
|
Brady™
76312 |
Black on Yellow | 15 ft. x 3 in. | Stripes |
Each for $82.44
|
|
|||
|
19017257
|
Brady™
78985 |
White | 15 ft. x 2 in. | Solid |
Each for $18.62
|
|
|||
|
19017258
|
Brady™
78986 |
Yellow | Solid |
Each for $20.30
|
|
||||
|
19017259
|
Brady™
78987 |
White | 15 ft. x 3 in. | Solid |
Each for $44.40
|
|
|||
|
19807082
|
Brady™
55314 |
Magenta on Yellow | 54 ft. x 3 in. | Stripes |
Each for $107.96
|
|
|||
|
19807242
|
Brady™
76316 |
Black on White | 54 ft. x 2 in. | Checkered |
Each for $76.55
|
|
|||
|
19807240
|
Brady™
76310 |
Black on White | Stripes |
Each for $48.39
|
|
||||
|
19807241
|
Brady™
76313 |
Red on White | Stripes |
Each for $82.44
|
|
||||
|
19807243
|
Brady™
76317 |
Black on Yellow | 54 ft. x 2 in. | Checkered |
Each for $77.41
|
|
|||
|
19807244
|
Brady™
76432 |
White Photoluminescent | 5 yd. x 2 in. | Solid |
Each for $126.62
|
|
|||
|
19807257
|
Brady™
78988 |
Yellow | 15 ft. x 3 in. | Solid |
Each for $48.39
|
|
|||
Description
- Ideal for temporary use outdoors or permanent indoor warning
- Sold as roll
B-957 Reflective Vinyl with Adhesive
- Thickness: 1.65 to 1.91mm (0.065 to 0.075 in.)
- Service temp.: -40° to 70°C (-40° to 160°F)
B-950 Encapsulated Vinyl with Adhesive
- Thickness: 0.15mm (0.006 in.)
- Service temp.: -40° to 80°C (-40° to 176°F)
Compliance
B-950 warning stripe and check tape meets OSHA specifications.