Learn More
Brady™ BradyBondz™ BMP71 Thermal Transfer Portable Printer Labels
Used to identify property from components to finished products
Supplier:  Brady™ M71 6 423
 View more versions of this product
Catalog No. 19000147
Only null left
Description
Description
  • White glossy polyester
  • Available in various sizes
Applications:
Asset Tracking, Barcoding, Component ID, Equipment ID, General Identification, General Purpose ID, Patch Panel - ID Port, Rating Plates, Through Hole - Top
Specifications
Chemical Identifiers
UL Recognised; CSA Accepted; AGA Approved; Halogen Free (DIN VDE 0472/part 815)
Permanent Acrylic
Gloss
BradyBondz Label
BMP™71 Portable Thermal Transfer Printer
-70°C to +110°C
BMP71 Label Printer Labels
White
Polyester (B-423)
Asset Tracking, Barcoding, Bottles, Component ID, Conical Tube, Deep Well Plate, Equipment ID, General Purpose ID, Petri Dish, Rating Plates, Through Hole - Top
Rectangle
0.076 mm
Specifications
UL Recognised; CSA Accepted; AGA Approved; Halogen Free (DIN VDE 0472/part 815)
Permanent Acrylic
Gloss
BradyBondz Label
BMP™71 Portable Thermal Transfer Printer
750/Roll
0.5 x 0.275 in.
-70°C to +110°C
10.16 mm
0.5 in.
0.275 in.
BMP71 Label Printer Labels
White
Polyester (B-423)
Asset Tracking, Barcoding, Bottles, Component ID, Conical Tube, Deep Well Plate, Equipment ID, General Purpose ID, Petri Dish, Rating Plates, Through Hole - Top
1
Rectangle
12.7 x 6.99 mm
0.076 mm
6.99 x 12.70 mm
12.70 mm
6.99 mm
Product Suggestions
Product Suggestions