Learn More
Brady™ BBP™ WARNING Pre-Printed Pre-Cut Blank Sign Headers
BBP™ WARNING Pre-Printed Pre-Cut Blank Sign Headers are durable, low-shrink vinyl with ultra-aggressive adhesive that is ideal for labeling pipes, equipment, shelves and nearly any other industrial surface.
$366.41 - $542.18
Specifications
Certifications/Compliance ASME ANSI Z535, OSHA 1910.145
Finish Gloss
For Use With (Equipment) BBP30, BBP31, BBP33, BBP35, and BBP37 printers
Material Vinyl
Adhesive Type Permanent Acrylic
View More Specs
Products 2
Catalog Number Mfr. No. Dimensions (L x W) Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Quantity Price Quantity & Availability  
19211126
View Documents Promotion Details
Brady™
B3025595ANSIWA
4 x 6 in. 175/Pk.
Each for $542.18
Only null left
19211125
View Documents Promotion Details
Brady™
B30241595ANSIWA
57 x 75 mm 300/Pk.
Each for $366.41
Only null left
Description
Description
  • Pre-printed headers result in multicolor header signs using a single, black print pass
  • Highly pliable material with high-tack permanent adhesive for rough, highly textured, dirty and harsh environment surfaces—including outdoors
  • Conforms and adheres to "difficult" surfaces like painted cinder blocks, painted wood, textured plastic, paper-jacketed pipes and blow-molded cases
  • Resistant to exposure to chemicals, greases and oils, salt spray, cleaners and detergents and general industrial grime
Specifications
Specifications
ASME ANSI Z535, OSHA 1910.145
BBP30, BBP31, BBP33, BBP35, and BBP37 printers
Permanent Acrylic
Pipemarkers, arrow tape, warehouse marking, warning panels, safety signs, arc flash and visual workplace labels
-40°F
-40°C to 82°C
English
Indoor/Outdoor Tape
Gloss
Vinyl
Pre-Printed Pre-Cut Blank Sign headers for BBP3x Printers
180°F
0.004 in.
-40°F to 180 °F
Warning
Black on Orange