Learn More
Brady™ BBP™ Retro Reflective Tape
Highly reflective identification that shines back brightly when struck by light
Supplier:  Brady™ B30C2250584OR
 View more versions of this product
Catalog No. 19211244
Only null left
Description
Description
  • Ideal for labeling outdoor utility poles and pedestals, jobsite equipment and roof-top equipment
  • Recommended low light or inclement weather conditions
  • Available in multiple colors
Specifications
Chemical Identifiers
Acrylic, Pressure-sensitive
Polyester
BBP30, BBP31, BBP33, BBP35, and BBP37 printers
4 to 6 Years
-40 to 158°F
0.0055 in.
15.24 m
Tape
For labeling outdoor utility poles and pedestals, jobsite equipment, roof-top equipment
100 Gardner Units
1 Roll
-40°C to +70°C
50 ft.
Specifications
Retro Reflective Tape for BBP3x Printers
Orange
Tape
For labeling outdoor utility poles and pedestals, jobsite equipment, roof-top equipment
100 Gardner Units
4 to 6 Years
Brady Workstation, LabelMark, MarkWare
-40°C to +70°C
0.0055 in.
50 ft.
2.25 in.
B30C-2250-584-OR
Acrylic, Pressure-sensitive
Gloss, Reflective
Polyester
BBP30, BBP31, BBP33, BBP35, and BBP37 printers
1
1 Roll
-40 to 158°F
10 lb./in.
0.14 mm
15.24 m
57.15 mm
Product Suggestions
Product Suggestions