Learn More
Brady™ BBP™ CAUTION Pre-Printed Pre-Cut Blank Sign Headers
BBP™ CAUTION Pre-Printed Pre-Cut Blank Sign Headers are made of durable, low-shrink vinyl with ultra-aggressive adhesive that is ideal for labeling pipes, equipment, shelves and nearly any other industrial surface.
Supplier:  Brady™ B30241595OSHACA
 View more versions of this product
Catalog No. 19211123
Only null left
Description
Description
  • Pre-printed headers result in multicolor header signs using a single, black print pass
  • Highly pliable material with high-tack permanent adhesive for rough, highly textured, dirty and harsh environment surfaces—including outdoors
  • Conforms and adheres to "difficult" surfaces like painted cinder blocks, painted wood, textured plastic, paper-jacketed pipes and blow-molded cases
  • Resistant to exposure to chemicals, greases and oils, salt spray, cleaners and detergents and general industrial grime
Specifications
Chemical Identifiers
ASME ANSI Z535, OSHA 1910.145
Vinyl
Pipemarkers, arrow tape, warehouse marking, warning panels, safety signs, arc flash and visual workplace labels
-40°F
-40°C to 82°C
CAUTION
BBP30, BBP31, BBP33, BBP35, and BBP37 printers
Permanent Acrylic
180°F
0.004 in.
-40°F to 180 °F
Indoor/Outdoor Tape
Specifications
ASME ANSI Z535, OSHA 1910.145
BBP30, BBP31, BBP33, BBP35, and BBP37 printers
300/Pk.
Pipemarkers, arrow tape, warehouse marking, warning panels, safety signs, arc flash and visual workplace labels
180°F
0.004 in.
76.2 mm
-40°F to 180 °F
CAUTION
Black on Yellow
B30-241-595-OSHACA
Vinyl
Permanent Acrylic
2.25 in.
-40°F
3 in.
-40°C to 82°C
2.25 x 3 in.
Indoor/Outdoor Tape
Product Suggestions
Product Suggestions