Learn More
Brady™ Aisle Marking Tapes
Ultra strong B-726 abrasion-resistant vinyl Aisle Marking Tape makes your facility safer, more visual and more efficient by adding colorful visual indicators to aisles and passageways.
Supplier:  Brady™ 58221
 View more versions of this product
Catalog No. 19046624
Only null left
Description
Description
  • Pressure sensitive rubber based adhesive sticks tightly to virtually any clean, dry surface
  • Good chemical, oil and water resistance
  • Ideal for marking aisles, passageways and storage locations and identifying equipment and egress paths
  • Quick application
Specifications
Chemical Identifiers
Acrylic
Vinyl
For pipe markers, arrow tape, warehouse marking, floor marking, warning panels, safety signs and hazardous substance identification
-4°F to 167°F
0.004 in.
33 m
Tape
Non-sterile
3 Years
-20°C to +75°C
36 yd.
Specifications
Acrylic
Tape
Marking Tape
For pipe markers, arrow tape, warehouse marking, floor marking, warning panels, safety signs and hazardous substance identification
1 Roll
-20°C to +75°C
36 yd.
3 in.
Blue
Vinyl
Non-sterile
3 Years
-4°F to 167°F
0.004 in.
33 m
76.2 mm
Product Suggestions
Product Suggestions