missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Aisle Marking Tapes

Catalog No. p-3697275
Change view
Click to view available options
Color:
Black
Blue
Green
Red
White
Yellow
Width (English):
2 in.
3 in.
Quantity:
1 Ea.
1 Roll
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Width (English) Quantity
19016206 Green 2 in. 1 Roll
19046622 Red 3 in. 1 Roll
19046623 Green 3 in. 1 Roll
19016204 Yellow 2 in. 1 Roll
19016213 White 3 in. 1 Roll
19046621 Black 2 in. 1 Ea.
19046625 Black 3 in. 1 Ea.
19046624 Blue 3 in. 1 Roll
19081380 Blue 2 in. 1 Roll
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 19016206 Supplier Brady™ Supplier No. 58202
Only null left

Ultra strong B-726 abrasion-resistant vinyl Aisle Marking Tape makes your facility safer, more visual and more efficient by adding colorful visual indicators to aisles and passageways.

  • Pressure sensitive rubber based adhesive sticks tightly to virtually any clean, dry surface
  • Good chemical, oil and water resistance
  • Ideal for marking aisles, passageways and storage locations and identifying equipment and egress paths
  • Quick application

Specifications

Adhesive Type Acrylic
Color Green
Format Tape
Material Vinyl
Product Type Marking Tape
Sterility Non-sterile
For Use With (Application) For pipe markers, arrow tape, warehouse marking, floor marking, warning panels, safety signs and hazardous substance identification
Life Span 3 Years
Quantity 1 Roll
Temperature Range (English) -4°F to 167°F
Temperature Range (Metric) -20°C to +75°C
Thickness (English) 0.004 in.
Length (English) 36 yd.
Length (Metric) 33 m
Width (English) 2 in.
Width (Metric) 50.8 mm
Show More Show Less